Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001346058.1 | NP_001332987.1 | 695 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris |