Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_548077.6 | XP_548077.2 | 326 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris |