Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005616353.3 | XP_005616410.1 | 3 - 4 | 454 | 65 | |
XM_005616354.3 | XP_005616411.1 | 3 - 4 | 578 | 65 | ||
XM_014119196.2 | XP_013974671.1 | 3 - 4 | 497 | 65 | ||
XM_014119197.2 | XP_013974672.1 | 3 - 4 | 563 | 65 | ||
XM_014119199.2 | XP_013974674.1 | 3 - 4 | 509 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Probe spans exons |