Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_845119.5 | XP_850212.2 | 2 - 3 | 567 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris Probe spans exons |