Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGES2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001131050.1 | NP_001124522.1 | 2 - 3 | 564 | 59 | |
XM_005625033.3 | XP_005625090.1 | 2 - 3 | 405 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |