Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD1LG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022421848.1 | XP_022277556.1 | 5 - 6 | 969 | 63 | |
XM_022421859.1 | XP_022277567.1 | 5 - 6 | 821 | 63 | ||
XM_847012.5 | XP_852105.3 | 5 - 6 | 964 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons |