Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B2M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003640047.2 | XP_003640095.1 | 2 - 3 | 567 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Probe spans exons |