Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022421263.1 | XP_022276971.1 | 6 - 7 | 1329 | 81 | |
XM_022421264.1 | XP_022276972.1 | 6 - 7 | 1331 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |