Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARFRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_850271.3 | XP_855364.2 | 4 - 5 | 404 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons |