Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHNAK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014121013.2 | XP_013976488.1 | 2 - 3 | 273 | 84 | |
XM_014121014.2 | XP_013976489.1 | 2 - 3 | 274 | 84 | ||
XM_022405790.1 | XP_022261498.1 | 2 - 3 | 273 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Probe spans exons |