Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639976.4 | XP_003640024.1 | 2 - 3 | 195 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |