Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014115189.2 | XP_013970664.1 | 3 - 4 | 521 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |