Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_849583.6 | XP_854676.3 | 4 - 5 | 493 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |