Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_848963.5 | XP_854056.1 | 3 - 4 | 302 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Canis familiaris Probe spans exons |