Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCUN1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005639754.3 | XP_005639811.1 | 5 - 6 | 748 | 146 | |
XM_014110427.2 | XP_013965902.1 | 5 - 6 | 629 | 146 | ||
XM_014110428.2 | XP_013965903.1 | 5 - 6 | 743 | 146 | ||
XM_843562.5 | XP_848655.2 | 5 - 6 | 734 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Probe spans exons |