Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637498.3 | XP_005637555.1 | 2 - 3 | 148 | 87 | |
XM_005637499.3 | XP_005637556.1 | 2 - 3 | 156 | 87 | ||
XM_022411903.1 | XP_022267611.1 | 2 - 3 | 148 | 87 | ||
XM_851633.5 | XP_856726.2 | 2 - 3 | 148 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |