Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUC7L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001131056.1 | NP_001124528.1 | 524 | 90 | ||
XM_005629474.2 | XP_005629531.1 | 503 | 90 | |||
XM_005629475.3 | XP_005629532.1 | 270 | 90 | |||
XM_005629477.3 | XP_005629534.1 | 847 | 90 | |||
XM_005629478.3 | XP_005629535.1 | 115 | 90 | |||
XM_014119776.2 | XP_013975251.1 | 278 | 90 | |||
XM_022403592.1 | XP_022259300.1 | 1006 | 90 | |||
XM_022403593.1 | XP_022259301.1 | 551 | 90 | |||
XM_022403595.1 | XP_022259303.1 | 577 | 90 | |||
XM_022403596.1 | XP_022259304.1 | 302 | 90 | |||
XM_022403597.1 | XP_022259305.1 | 180 | 90 | |||
XM_022403600.1 | XP_022259308.1 | 172 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris |