Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076794.1 | NP_001070262.1 | 36 - 37 | 5570 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Canis familiaris Probe spans exons |