Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252264.1 | NP_001239193.1 | 1 | 349 | 113 | |
XM_005632022.3 | XP_005632079.1 | 1 | 345 | 113 | ||
XM_022406218.1 | XP_022261926.1 | 1 | 362 | 113 | ||
GenBank mRNA | DN371621.1 | - | 304 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |