Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022409133.1 | XP_022264841.1 | 9 - 10 | 1113 | 93 | |
XM_542952.6 | XP_542952.3 | 10 - 11 | 1278 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |