Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIPK2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_850186.5 | XP_855279.2 | 4 | 1223 | 174 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Amplicon spans exons and probe does not span exons |