Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGFRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005618675.3 | XP_005618732.2 | 8 | 1959 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |