Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005624948.3 | XP_005625005.1 | 15 - 16 | 3712 | 105 | |
XM_548319.6 | XP_548319.3 | 15 - 16 | 3758 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |