Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014110679.1 | XP_013966154.1 | 8 | 2914 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |