Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRSF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639860.3 | XP_003639908.1 | 3 - 4 | 438 | 151 | |
XM_005635077.2 | XP_005635134.1 | 3 - 4 | 1051 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |