Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFAP2E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014119246.2 | XP_013974721.1 | 3 | 352 | 182 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Amplicon spans exons and probe does not span exons |