Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005624787.3 | XP_005624844.1 | 3 | 895 | 152 | |
XM_022423280.1 | XP_022278988.1 | 4 | 1027 | 152 | ||
XM_022423281.1 | XP_022278989.1 | 4 | 1010 | 152 | ||
XM_022423282.1 | XP_022278990.1 | 3 | 435 | 152 | ||
XM_548251.6 | XP_548251.1 | 3 | 915 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |