Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008552.1 | NP_001008552.1 | 552 | 119 | ||
XM_014121534.2 | XP_013977009.1 | 701 | 119 | |||
GenBank mRNA | AY764285.1 | - | 552 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Inventoried |