Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005623417.3 | XP_005623474.1 | 1 | 428 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |