Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005627482.3 | XP_005627539.1 | 3 | 1380 | 79 | |
XM_022426159.1 | XP_022281867.1 | 2 | 1440 | 79 | ||
XM_538967.6 | XP_538967.2 | 2 | 1479 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Amplicon spans exons and probe does not span exons |