Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP51A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005628499.3 | XP_005628556.1 | 6 - 7 | 838 | 196 | |
XM_022427338.1 | XP_022283046.1 | 6 - 7 | 812 | 196 | ||
XM_532457.6 | XP_532457.3 | 6 - 7 | 1041 | 196 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Probe spans exons |