Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMBS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005619681.3 | XP_005619738.1 | 1032 | 148 | ||
XM_005619682.3 | XP_005619739.1 | 933 | 148 | |||
XM_005619683.3 | XP_005619740.1 | 1093 | 148 | |||
XM_014113375.2 | XP_013968850.1 | 1277 | 148 | |||
XM_022418216.1 | XP_022273924.1 | 957 | 148 | |||
XM_546491.6 | XP_546491.2 | 946 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Endogenous Controls Canis familiaris |