Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165888.1 | NP_001159360.1 | 4 | 146 | ||
XM_005633296.3 | XP_005633353.1 | 5 | 146 | |||
GenBank mRNA | FN395288.1 | - | 3 | 881 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |