Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_845177.5 | XP_850270.1 | 2 - 3 | 609 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |