Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTHFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005617992.3 | XP_005618049.1 | 10 - 11 | 1986 | 181 | |
XM_005617993.3 | XP_005618050.1 | 10 - 11 | 1768 | 181 | ||
XM_014111461.2 | XP_013966936.1 | 11 - 12 | 1730 | 181 | ||
XM_014111464.2 | XP_013966939.1 | 9 - 10 | 1539 | 181 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons |