Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUV39H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_548987.6 | XP_548987.2 | 4 - 5 | 1101 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons |