Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK5R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005624820.3 | XP_005624877.1 | 1 | 266 | 74 | |
XM_022423313.1 | XP_022279021.1 | 1 | 266 | 74 | ||
XM_022423314.1 | XP_022279022.1 | 1 | 266 | 74 | ||
XM_022423315.1 | XP_022279023.1 | 1 | 266 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |