Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISLR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005638599.3 | XP_005638656.2 | 2 | 1229 | 156 | |
XM_005638600.3 | XP_005638657.1 | 2 | 1398 | 156 | ||
XM_022412662.1 | XP_022268370.1 | 2 | 1323 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |