Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005640502.3 | XP_005640559.1 | 5 - 6 | 2949 | 97 | |
XM_536020.6 | XP_536020.4 | 5 - 6 | 3130 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris Probe spans exons |