Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOX30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003434517.4 | XP_003434565.1 | 3 | 1615 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Amplicon spans exons and probe does not span exons |