Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHBDD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005636409.3 | XP_005636466.1 | 4 | 895 | 132 | |
XM_005636411.3 | XP_005636468.1 | 5 | 1051 | 132 | ||
XM_005636412.3 | XP_005636469.1 | 3 | 741 | 132 | ||
XM_005636413.2 | XP_005636470.1 | 3 | 654 | 132 | ||
XM_005636414.3 | XP_005636471.1 | 3 | 726 | 132 | ||
XM_022410477.1 | XP_022266185.1 | 4 | 1138 | 132 | ||
XM_022410478.1 | XP_022266186.1 | 4 | 886 | 132 | ||
XM_022410479.1 | XP_022266187.1 | 4 | 803 | 132 | ||
XM_022410480.1 | XP_022266188.1 | 3 | 977 | 132 | ||
XM_543467.5 | XP_543467.2 | 3 | 651 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |