Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BARHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_548404.5 | XP_548404.3 | 1 | 976 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Amplicon spans exons and probe does not span exons |