Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSHZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005616783.3 | XP_005616840.1 | 2 | 2686 | 109 | |
XM_022426433.1 | XP_022282141.1 | 3 | 2846 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |