Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHODL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005638776.3 | XP_005638833.1 | 2 | 307 | 134 | |
XM_014109868.2 | XP_013965343.1 | 2 | 557 | 134 | ||
XM_544823.6 | XP_544823.2 | 2 | 307 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |