Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCHF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_538609.6 | XP_538609.2 | 3 - 4 | 690 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Canis familiaris Probe spans exons |