Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005627618.3 | XP_005627675.1 | 2 | 414 | 144 | |
XM_005627619.3 | XP_005627676.1 | 3 | 497 | 144 | ||
XM_005627620.3 | XP_005627677.1 | 2 | 4443 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |