Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197185.1 | NP_001184114.1 | 1 - 2 | 531 | 131 | |
NM_001197186.1 | NP_001184115.1 | 1 - 2 | 531 | 131 | ||
XM_005641004.3 | XP_005641061.1 | 1 - 2 | 360 | 131 | ||
XM_005641005.3 | XP_005641062.1 | 1 - 2 | 360 | 131 | ||
XM_005641006.3 | XP_005641063.1 | 1 - 2 | 360 | 131 | ||
GenBank mRNA | AY530157.1 | - | 1 - 2 | 161 | 131 | |
AY530158.1 | - | 1 - 2 | 161 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |