Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_542970.6 | XP_542970.2 | 11 - 12 | 1234 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris Probe spans exons |