Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005626717.3 | XP_005626774.1 | 2 - 3 | 289 | 162 | |
XM_005626718.3 | XP_005626775.1 | 3 - 4 | 364 | 162 | ||
XM_531977.6 | XP_531977.1 | 2 - 3 | 271 | 162 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Probe spans exons |