Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197190.1 | NP_001184119.1 | 3 - 4 | 422 | 141 | |
GenBank mRNA | DN373942.1 | - | 3 - 4 | 430 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Probe spans exons |